Learn More
Alkali Scientific 15mL Centrifuge Tubes, PP (17 X 118mm), Flat Screw Cap, Individually Wrapped, Sterile, 300/cs
SDP
Catalog No. 502389574
Encompass_Preferred
15mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free
Catalog No. 50-238-9574 Supplier Alkali Scientific Supplier No. C5615W
15mL Centrifuge Tubes: Conical centrifuge tubes, high clarity polypropylene, printed graduation marks, sterile, certified RNase, DNase and pyrogen free