Learn More
CELLTREAT Scientific Products 15mL Centrifuge Tube, Sure Cap - Bag, Sterile
SDP
Catalog No. 501117982
Encompass_Preferred
CELLTREAT Scientific Products 15mL Sure Cap Centrifuge Tube, Polypropylene. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps. Compressible liner creates a stable seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Tubes have a Max Force (RCF) g of 12000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 500/case.
Catalog No. 50-111-7982 Supplier CELLTREAT Scientific Products Supplier No. 229401
CELLTREAT Scientific Products 15mL Sure Cap Centrifuge Tube, Polypropylene. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps. Compressible liner creates a stable seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Tubes have a Max Force (RCF) g of 12000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 500/case.