Learn More
JDM LAB SOLUTIONS LLC 15mL ProSeries Tubes Sterile Bags
Catalog No. NC4039947
Encompass
ProSeries™ High Performance Centrifuge Tubes, 15mL, 20,000xg max RCF, sterile, w/ Hi-Viz Graduations and FreezeTech Temperature Indicator Patch, 20 eco-plastic racks of 25 tubes, 500/cs
Catalog No. NC4039947 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMC2801
ProSeries™ High Performance Centrifuge Tubes, 15mL, 20,000xg max RCF, sterile, w/ Hi-Viz Graduations and FreezeTech Temperature Indicator Patch, 20 eco-plastic racks of 25 tubes, 500/cs