Learn More
Labcon North America Dry Ice Storage Tubes, 15 mL, 10/pk
Catalog No. NC1704298
Encompass_Preferred
15mL ProtectR Dry Ice Storage Tubes in IntegraPack, Sterile, 10/pk.
Catalog No. NC1704298 Supplier Labcon North America Supplier No. 38303250009
15mL ProtectR Dry Ice Storage Tubes in IntegraPack, Sterile, 10/pk.