Learn More
CELLTREAT Scientific Products 15mL Polystyrene Centrifuge Tube, Sure Cap - Bag, Sterile
SDP
Catalog No. 501117983
Encompass_Preferred
CELLTREAT Scientific Products 15mL Sure Cap Centrifuge Tube, Polystyrene. Easy to write on flat surface polyethylene (HDPE) caps. Compressible liner creates a stable seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Polystyrene is optically clear for easy viewing of sample or colormetric testing. Note: Polystyrene is less chemically compatible than polypropylene, and can not withstand high speed centrifugation. For a polypropylene option, see 229401. Max Force (RCF) g 3000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable Bag, 500/case.
Catalog No. 50-111-7983 Supplier CELLTREAT Scientific Products Supplier No. 229491
CELLTREAT Scientific Products 15mL Sure Cap Centrifuge Tube, Polystyrene. Easy to write on flat surface polyethylene (HDPE) caps. Compressible liner creates a stable seal. Tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient zip-closure bag is a practical alternative to folding and taping opened bags. Polystyrene is optically clear for easy viewing of sample or colormetric testing. Note: Polystyrene is less chemically compatible than polypropylene, and can not withstand high speed centrifugation. For a polypropylene option, see 229401. Max Force (RCF) g 3000. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable Bag, 500/case.