Learn More
Sarstedt Inc 15mL Screw Cap Tube, 17mm Diameter x 120mm Long, 500/Case
Catalog No. NC9859685 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube, Reagent and Centrifuge; 15mL; Polystyrene; 120L x 17mm dia.; Conical Base; Transparent; Non-sterile; Red Screw Cap; Max. Centrifugation Speed: 4100 x g Centrifuge with cone adapter; With printed writing space and graduation; 500/CS
Catalog No. NC9859685 Supplier Sarstedt Inc Supplier No. 62.553.001
Tube, Reagent and Centrifuge; 15mL; Polystyrene; 120L x 17mm dia.; Conical Base; Transparent; Non-sterile; Red Screw Cap; Max. Centrifugation Speed: 4100 x g Centrifuge with cone adapter; With printed writing space and graduation; 500/CS