Learn More
Sarstedt Inc 15ML SC TUBE 20X76 CBS S/PK500
Supplier:  Sarstedt Inc 60.732.001
Encompass_Preferred
15ml Screw Cap Tube 20x76mm Conical Bottom Skirted Polypropylene Cap Assembled Sterile
Catalog No. NC9374357