Learn More
Supply Solutions 15mL Screw Cap PYREX Culture Tubes with PTFE Lined Phenolic Caps 16x125mm
Catalog No. NC3052580
Encompass_Preferred
15mL Screw Cap PYREX Culture Tubes with PTFE Lined Phenolic Caps 16x125mm
Catalog No. NC3052580 Supplier Supply Solutions Supplier No. 982616X
15mL Screw Cap PYREX Culture Tubes with PTFE Lined Phenolic Caps 16x125mm
Can these glass products handle heat and chemicals?
Yes, Type I, Class A borosilicate glass is designed for excellent heat resistance and is highly resistant to chemical attack, making it suitable for various laboratory applications.
How should borosilicate glassware be handled for optimal durability?
Proper care ensures the glassware withstands most laboratory temperatures and maintains its chemical resistance. The mechanical strength depends not only on wall thickness but also on the glass formula and type.
What makes Type I, Class A borosilicate glass ideal for laboratory applications?
• Temperature Resistance: Withstands nearly all temperatures used in normal laboratory use with proper care
• Chemical Resistance: Highly resistant to chemical attack
• Low Coefficient of Expansion: Allows for the production of glass with varying wall thicknesses tailored for specific purposes
• Durability: Mechanical strength is influenced by the glass formula and type, ensuring reliability and heat resistance
What are the key features of the materials used in these products?
• Non-pyrogenic
• Human DNA-free
• PCR inhibitor-free
• RNase-/DNase-free
• Made from USP Class VI Polypropylene (for plastic components)