Learn More
Sarstedt Inc 15mL Screen Printed Screw Cap Tube, Conical Bottom, Polystyrene, 500/Case
Catalog No. 50809219 Shop All Sarstedt Inc. Products
Encompass_Preferred
15mL Screen Printed Screw Cap Tube 17x120mm Conical Bottom Polystyrene Graduations and Writing Block Rack Pack Red Cap Assembled Sterile
Catalog No. 50-809-219 Supplier Sarstedt Inc Supplier No. 62.553.002
15mL Screen Printed Screw Cap Tube 17x120mm Conical Bottom Polystyrene Graduations and Writing Block Rack Pack Red Cap Assembled Sterile