Learn More
Thermo Scientific 15ml Tube Block Set with Cover
Supplier:  Thermo Scientific 88880151
Encompass_Preferred
No. of Places 15, Tube Size 15mL, For use with Digital Heating Shaking Drybath 88880028, 88880027
Catalog No. NC1587746