Learn More
Chemglass Life Sciences 15mL Single Neck RBF, 19/22 Outer with Hooks
SDP
Catalog No. 5000980613
Encompass_Preferred
Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint. Flask has two hooks 180 degrees apart for use with CG-110 stainless steel springs. Flasks with joint sizes other than those listed are available, please call for pricing.
Catalog No. 50-009-80613 Supplier Chemglass Life Sciences Supplier No. CG150706
Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint. Flask has two hooks 180 degrees apart for use with CG-110 stainless steel springs. Flasks with joint sizes other than those listed are available, please call for pricing.