Learn More
Alkali Scientific ASI™ 15mL Skirted Centrifuge Tube, Self Standing, Snap Cap, PP/HDPE, Sterile, Non-Pyrogenic, Non-Cytotoxic, DNase/RNase Free, 500/Cs
SDP
Catalog No. 503029238
Encompass_Preferred
Snap tubes are widely used in laboratories for multiple purposes, from storage of samples to high speed centrifugation. Snap tubes can assist users during experiments by enabling easy one-hand opening and closing of the tubes by providing snap type cap. ▪ Snap cap with tight sealing▪ High chemical resistance▪ Recommended volume for centrifugation: 80%▪ Imprinted external graduations with marking area ▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-9238 Supplier Alkali Scientific Supplier No. CSP5022
Snap tubes are widely used in laboratories for multiple purposes, from storage of samples to high speed centrifugation. Snap tubes can assist users during experiments by enabling easy one-hand opening and closing of the tubes by providing snap type cap. ▪ Snap cap with tight sealing▪ High chemical resistance▪ Recommended volume for centrifugation: 80%▪ Imprinted external graduations with marking area ▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free