Learn More
Cole-Parmer 15 mL PFA Vials in organizer trays, 10/tray
Supplier:  Cole-Parmer 0893630
Encompass_Preferred
PFA Vials in Organizer Trays are inert and nonleaching, with a temperature range of –320 to 500°F (–195 to 260°C)—ideal for sensitive samples. Trays are black HDPE with numbered pockets and slots for identification cards (not included). Conical vial facilitates removal of contents with a syringe. The 7-mL vials have a 6-7⁄8"L x 2-3⁄4"W (17.5 x 7.0 cm) tray; the 15-, 22-, and 30-mL vials have a 10-1⁄2"L x 3-1⁄4"W (26.7 x 8.3 cm) tray. Each tray contains ten vials.
Catalog No. NC1084632