Learn More
Ace Glass, Inc. 15mL tissue grinder consists of all-glass pestle and tapered tube
SDP
Catalog No. 50974569
Encompass_Preferred
15mL tissue grinder consists of all-glass pestle and tapered tube. The conical area is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1 - 0.15mm clearance. The 10mm pestle rod can be used with a 509558293 Flex-Grip Chuck. Capacity represents volume with pestle inserted.
This item is not returnable. View return policy
Catalog No. 50-974-569 Supplier Ace Glass, Inc. Supplier No. 832520
15mL tissue grinder consists of all-glass pestle and tapered tube. The conical area is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1 - 0.15mm clearance. The 10mm pestle rod can be used with a 509558293 Flex-Grip Chuck. Capacity represents volume with pestle inserted.