Learn More
Diagenode Inc 15ML TPX TUBES (50PCS)
Catalog No. NC0150935
Encompass
15 Ml Bioruptor Plus Tpx Tubes, 50 Pc Format. Tpx Polymethylpentene (Pmp) Tubes Improve Sonication And Shearing Efficiency. Tpx Is A Hard Plastic Which Ameliorates The Transmission Of The Ultrasound To The Sample. Tpx Tubes Have Been Validated For Use On The Bioruptors Standard And Bioruptors Plus. Recommended Sample Volume: 500 Ul - 2 Ml.
Catalog No. NC0150935 Supplier Diagenode Inc Supplier No. C30010009
15 Ml Bioruptor Plus Tpx Tubes, 50 Pc Format. Tpx Polymethylpentene (Pmp) Tubes Improve Sonication And Shearing Efficiency. Tpx Is A Hard Plastic Which Ameliorates The Transmission Of The Ultrasound To The Sample. Tpx Tubes Have Been Validated For Use On The Bioruptors Standard And Bioruptors Plus. Recommended Sample Volume: 500 Ul - 2 Ml.