Learn More
CELLTREAT Scientific Products 15mL Tube Cell Strainer, 100μm, Yellow, Individually Wrapped, Sterile
SDP
Catalog No. 5000144339
Encompass_Preferred
Versatile design of 15mL Cell Strainers accommodates a wide range of tubes, from 0.5mL Micro Tubes through 15mL Centrifuge TubesStackable design allows for serial filtrationCan be used for many applications including isolating cells and removing clumps or debrisUpper cup can be removed and inverted to collect residual cellsHydrophilic treatment and venting spacers increase flow and reduce the effects of surface tensionPolypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingExclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers
Catalog No. 50-001-44339 Supplier CELLTREAT Scientific Products Supplier No. 230475
Versatile design of 15mL Cell Strainers accommodates a wide range of tubes, from 0.5mL Micro Tubes through 15mL Centrifuge TubesStackable design allows for serial filtrationCan be used for many applications including isolating cells and removing clumps or debrisUpper cup can be removed and inverted to collect residual cellsHydrophilic treatment and venting spacers increase flow and reduce the effects of surface tensionPolypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materialsExtended lip on housing enables aseptic handling with forcepsElongated gripping tab on housing for easy handlingExclusive pestle (229480) is an inexpensive and convenient tool for use with Cell Strainers