Learn More
Ace Glass, Inc. 15mL volumetric bulb for use with ACE Automatic Dispensing Pipet
SDP
Catalog No. 50975873
Encompass_Preferred
15mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-975-873 Supplier Ace Glass, Inc. Supplier No. 800430
15mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.