Learn More
Ace Glass, Inc. 15mm Ace-Thred PTFE bushing, -110 FETFE O-ring, accepts12.5-14mm OD tubing
SDP
Supplier:  Ace Glass, Inc. 750627
Encompass_Preferred
15mm Ace-Thred PTFE bushing connector for joining an Ace-Thred to a reduced-end tube. Bushing accepts 12.5-14mm O.D. tubing. Includes -110 FETFE o-ring.
Catalog No. 50-955-7816