Learn More
Ace Glass, Inc. 15MM GRADED SEALS 15MM GRADED SEALS
SDP
Catalog No. 50979782
Encompass
15mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-782 Supplier Ace Glass, Inc. Supplier No. 845524
15mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.