Learn More
Ace Glass, Inc. 15mm adapter, nylon, polyethylene packing support, Luer slip bottom drip
SDP
Catalog No. 50972238
Encompass_Preferred
15mm bottom-drip, Luer-Lok, nylon adapter with flow regulator valve for use with all #15 Ace- Thred chromatography columns. Bore is adjustable from 0-1mm max. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-972-238 Supplier Ace Glass, Inc. Supplier No. 583711
15mm bottom-drip, Luer-Lok, nylon adapter with flow regulator valve for use with all #15 Ace- Thred chromatography columns. Bore is adjustable from 0-1mm max. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.