Learn More
Ace Glass, Inc. #15 Ace-Thred solid nylon plug, 7855-716 FETFE O-ring front seal
SDP
Catalog No. 509557161
Encompass_Preferred
#15 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -110 FETFE O-ring.
Catalog No. 50-955-7161 Supplier Ace Glass, Inc. Supplier No. 584612
#15 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -110 FETFE O-ring.