Learn More
Chemglass Life Sciences 15mm Quartz Fritted Disc, Medium
SDP
Catalog No. CGQ020704
Encompass_Preferred
15mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020704 Supplier Chemglass Life Sciences Supplier No. CGQ020704
15mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.