Learn More
Bellco Glass, Inc. 15mm Round Cover Glass
SDP
Catalog No. 502329610
Encompass_Preferred
High-quality, optically clear borosilicate glass cover slips are ideal for specialty microscopy and imagingIdeal for use with dishes and multiple well plates and anywhere a square cover glass is unsuitableConsistent flatness and scratch-freeResistant to organic solventsPackaged in a hinged plastic storage case15mm Round Cover Glass can be used with a 24 well glass bottom plate (Part No. 229125)Cover Glass is #1.5 thickness Pack Description100/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialGlassTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-232-9610 Supplier Bellco Glass, Inc. Supplier No. 229172000
High-quality, optically clear borosilicate glass cover slips are ideal for specialty microscopy and imagingIdeal for use with dishes and multiple well plates and anywhere a square cover glass is unsuitableConsistent flatness and scratch-freeResistant to organic solventsPackaged in a hinged plastic storage case15mm Round Cover Glass can be used with a 24 well glass bottom plate (Part No. 229125)Cover Glass is #1.5 thickness Pack Description100/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialGlassTissue Culture TreatedNoRNase/DNase FreeYes