Learn More
Ace Glass, Inc. 15mm adapter, PTFE, polyethylene packing support, Luer slip bottom drip with flow regulator valve
SDP
Catalog No. 50978488
Encompass_Preferred
25mm bottom-drip, Luer-Lok, PTFE adapter for use with all #25 Ace- Thred chromatography columns. Bore is 1mm. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.
Catalog No. 50-978-488 Supplier Ace Glass, Inc. Supplier No. 583753
25mm bottom-drip, Luer-Lok, PTFE adapter for use with all #25 Ace- Thred chromatography columns. Bore is 1mm. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.