Learn More
Med Vet International 15mm x 32mm Cuttable Titanium TTA Cage, J1072SCTC
Catalog No. 5000583740
Encompass_Preferred
As additional sizes of cages become avail- able it is becoming increasingly difficult to manage and store the full inventory of cages. The expense of keeping the current 24 cage sizes in stock is considerable. Use of cuttable cages means that less cages need to be stocked (particularly the short uncommon sizes), cage length can be adjusted quickly and easily for each patient during the procedure. Cages may be trimmed by 2mm increments using the dedicated cutters..Cuttable TTA Cage Titanium 15mm x 32mm
Catalog No. 50-005-83740 Supplier Med Vet International Supplier No. JOR1072SCTC
As additional sizes of cages become avail- able it is becoming increasingly difficult to manage and store the full inventory of cages. The expense of keeping the current 24 cage sizes in stock is considerable. Use of cuttable cages means that less cages need to be stocked (particularly the short uncommon sizes), cage length can be adjusted quickly and easily for each patient during the procedure. Cages may be trimmed by 2mm increments using the dedicated cutters..Cuttable TTA Cage Titanium 15mm x 32mm