Learn More
Cayman Chemical 15mthyl PalmItIc ACD mthy 10mg
SDP
Catalog No. 503016003
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in S. zeai sea sponges as the fatty acyl component of zeamide, oolong tea, and P. sulcate extract
Catalog No. 50-301-6003 Supplier Cayman Chemical Supplier No. 2481510 MG
A methylated fatty acid methyl ester; has been found in S. zeai sea sponges as the fatty acyl component of zeamide, oolong tea, and P. sulcate extract