Learn More
Cayman Chemical 15PGDH ngatIve Control15 250uL
SDP
Catalog No. 502893285
Encompass_Preferred
An a1-adrenergic and 5-HT2A receptor antagonist (IC50s 2A- and a2B-adrenergic and 5-HT1A 5-HT1B 5-HT2B and 5-HT2C receptors (IC50s 18.1 4.89 4.68 41 11.5 and 26 nM respectively) among others decreases systolic and diastolic blood pressure and heart rate in spontaneously hypertensive rats and 2K-1C renal hypertensive dogs at 10 and 4 mg/kg respectively
Catalog No. 50-289-3285 Supplier Cayman Chemical Supplier No. 701984250 UL
An a1-adrenergic and 5-HT2A receptor antagonist (IC50s 2A- and a2B-adrenergic and 5-HT1A 5-HT1B 5-HT2B and 5-HT2C receptors (IC50s 18.1 4.89 4.68 41 11.5 and 26 nM respectively) among others decreases systolic and diastolic blood pressure and heart rate in spontaneously hypertensive rats and 2K-1C renal hypertensive dogs at 10 and 4 mg/kg respectively