Learn More
USA Scientific Inc 15-place Upright freezer rack
Catalog No. 502745116
Encompass
15-place (3 long x 5 high) upright freezer rack for 100-place hinged boxes, 18 3/4 in L x 5 7/8 in W x 11 9/16 in H, stainless steel
Catalog No. 50-274-5116 Supplier USA Scientific Inc Supplier No. 26101500
15-place (3 long x 5 high) upright freezer rack for 100-place hinged boxes, 18 3/4 in L x 5 7/8 in W x 11 9/16 in H, stainless steel