Learn More
USA Scientific Inc 15-place upright freezer rk
Catalog No. 502744747
Encompass
15-place (3 long x 5 high) upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 11 in H, stainless steel
Catalog No. 50-274-4747 Supplier USA Scientific Inc Supplier No. 26023500
15-place (3 long x 5 high) upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 11 in H, stainless steel