Learn More
Enzo Life Sciences 15(S)-Hydroxyeicosa-5Z,8Z,11Z,13E-tetraenoic acid (HETE) (1mg). CAS: 54845-95-3
SDP
Catalog No. 502011020
Encompass_Preferred
Biosynthesis in human neutrophils, eosinophils and reticulocytes. Most abundant eicosanoid in human bronchi. Regulates T lymphocyte mitogenesis. Induces immunosupression. Stimulates migration of capillary endothelial cells and neovascularization. Alternative name: 15(S)-HETE. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1020 Supplier Enzo Life Sciences Supplier No. BMLH0151000
Add to Cart
Add to Cart
Biosynthesis in human neutrophils, eosinophils and reticulocytes. Most abundant eicosanoid in human bronchi. Regulates T lymphocyte mitogenesis. Induces immunosupression. Stimulates migration of capillary endothelial cells and neovascularization. Alternative name: 15(S)-HETE. Formulation: Oil dissolved in ethanol. Purity: ≥99% (HPLC). Long Term Storage: -80°C.