Learn More
Bulbman Inc F15T8CW 15W 18IN FLUORESCENT
SDP
Catalog No. NC9000880
Encompass_Preferred
15 Watt Fluorescent Lamp - Cool White 4100 Kelvin - G13 Bi-pin Base - 18 Inch Length
Catalog No. NC9000880 Supplier Bulbman Inc Supplier No. 112591
15 Watt Fluorescent Lamp - Cool White 4100 Kelvin - G13 Bi-pin Base - 18 Inch Length