Learn More
Chemglass Life Sciences 15 Well Evaporator/Concentrator Manifold, Replaceable Needles
SDP
Catalog No. 507513687
Encompass_Preferred
15 Well Evaporator/Concentrator Manifold, Replaceable Needles. Manifold is a low cost solution for the concentration of product in 96, 48 or 24 well blocks. Delivers equal amounts of gas to each well to gently speed solvent evaporation. Needles are constructed of stainless steel. The 15 well units have removable/replaceable needles. Manifold is constructed of polypropylene and the base is coated aluminum. Equipped with an adjustable flow meter for controlling gas flow into the manifold. Inlet is a 1/8” barbed fitting.Note: Blocks are not included and must be purchased separately.
Catalog No. 50-751-3687 Supplier Chemglass Life Sciences Supplier No. OP502007
15 Well Evaporator/Concentrator Manifold, Replaceable Needles. Manifold is a low cost solution for the concentration of product in 96, 48 or 24 well blocks. Delivers equal amounts of gas to each well to gently speed solvent evaporation. Needles are constructed of stainless steel. The 15 well units have removable/replaceable needles. Manifold is constructed of polypropylene and the base is coated aluminum. Equipped with an adjustable flow meter for controlling gas flow into the manifold. Inlet is a 1/8” barbed fitting.Note: Blocks are not included and must be purchased separately.