Learn More
Chemglass Life Sciences Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 15 x 1.5
SDP
Catalog No. CG200318
Encompass_Preferred
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 15 x 1.5. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2003-50 FOR STIR BAR KIT
Catalog No. CG200318 Supplier Chemglass Life Sciences Supplier No. CG200318
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 15 x 1.5. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2003-50 FOR STIR BAR KIT