Learn More
S2 Media 15x100mm Aspergillus Differential Agar, 10/pack
Catalog No. 502995778
Encompass_Preferred
Aspergillus Differential Agar is a non-selective, differential medium used for the presumptive identification of Aspergillus flavus. In 1974, Bothast and Fennell developed this formulation as a screening medium, used to detect Aspergillus flavus and closely related species. Any isolate of Aspergillus of clinical significance should be identified. • Product meets BAM, CLSI, ISO, and SMWW reference methods and performance criteria.• sMAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5778 Supplier S2 Media Supplier No. 5084
Aspergillus Differential Agar is a non-selective, differential medium used for the presumptive identification of Aspergillus flavus. In 1974, Bothast and Fennell developed this formulation as a screening medium, used to detect Aspergillus flavus and closely related species. Any isolate of Aspergillus of clinical significance should be identified. • Product meets BAM, CLSI, ISO, and SMWW reference methods and performance criteria.• sMAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.