Learn More
S2 Media 15x100mm Baird Parker (BP) Agar, 10/pack
Catalog No. 502995874
Encompass_Preferred
Baird-Parker (BP) Agar is a selective and differential medium used for the isolation and presumptive identification of coagulase-positive Staphylococcus from clinical specimens, foods, air, water, and other materials. Colonies of S. aureus form a characteristic black pigmentation by reducing the potassium tellurite and form clear zones due to proteolysis of the egg yolk emulsion. Lecithinase or lipase production results in the formation of a smaller, opaque zone within the clear zone. • Product meets AOAC, BAM, CLSI, EP, ISO, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BP Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5874 Supplier S2 Media Supplier No. 5058
Baird-Parker (BP) Agar is a selective and differential medium used for the isolation and presumptive identification of coagulase-positive Staphylococcus from clinical specimens, foods, air, water, and other materials. Colonies of S. aureus form a characteristic black pigmentation by reducing the potassium tellurite and form clear zones due to proteolysis of the egg yolk emulsion. Lecithinase or lipase production results in the formation of a smaller, opaque zone within the clear zone. • Product meets AOAC, BAM, CLSI, EP, ISO, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BP Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.