Learn More
S2 Media 15x100mm Buffered Charcoal Yeast Extract Agar with Polymyxin B, Anisomycin, and Cefamandole (BCYE-PAC), Deep Fill, 10/pack
Catalog No. 502995910
Encompass_Preferred
Buffered Charcoal Yeast Extract Agar with Polymyxin B, Anisomycin, and Cefamandole (BCYE-PAC) is a selective, non-differential medium for the isolation of Legionella species. L-cysteine HCl, ferric pyrophosphate and alpha-ketoglutarate provide nutrients required by Legionella species. Activated charcoal decomposes hydrogen peroxide, which is a toxic metabolic biproduct, while also collecting CO2 and modifying surface tension of the media. ACES buffer helps maintain the proper pH for optimal growth. Polymyxin B, Anisomycin, and cefamandole are selective agents added to suppress background flora present in samples. • Product meets CLSI, EPA, CDC, ISO, and SMWW reference methods and performance criteria.• BCYE-PAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5910 Supplier S2 Media Supplier No. 5041
Buffered Charcoal Yeast Extract Agar with Polymyxin B, Anisomycin, and Cefamandole (BCYE-PAC) is a selective, non-differential medium for the isolation of Legionella species. L-cysteine HCl, ferric pyrophosphate and alpha-ketoglutarate provide nutrients required by Legionella species. Activated charcoal decomposes hydrogen peroxide, which is a toxic metabolic biproduct, while also collecting CO2 and modifying surface tension of the media. ACES buffer helps maintain the proper pH for optimal growth. Polymyxin B, Anisomycin, and cefamandole are selective agents added to suppress background flora present in samples. • Product meets CLSI, EPA, CDC, ISO, and SMWW reference methods and performance criteria.• BCYE-PAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.