Learn More
S2 Media 15x100mm Brain Heart Infusion (BHI) Agar with 7% Sheep Blood, Chloramphenicol, and Gentamicin, 10/pack
Catalog No. 502995759
Encompass_Preferred
Brain Heart Infusion (BHI) Agar supplemented with 7% Sheep Blood, Chloramphenicol, and Gentamicin produces a selective medium utilized for the isolation of pathogenic fungi from specimens contaminated with bacteria. Gentamicin inhibits the growth of gram-negative and some gram-positive bacteria. Chloramphenicol is a broad-spectrum antimicrobic that also inhibits a range of gram-positive and gram-negative bacteria. Sheep blood is used to enrich the base and support growth of systemic fungi that may grow poorly on nonenriched medium.• Product meets ASM and CLSI reference methods and performance criteria.• BHI should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5759 Supplier S2 Media Supplier No. 5115
Brain Heart Infusion (BHI) Agar supplemented with 7% Sheep Blood, Chloramphenicol, and Gentamicin produces a selective medium utilized for the isolation of pathogenic fungi from specimens contaminated with bacteria. Gentamicin inhibits the growth of gram-negative and some gram-positive bacteria. Chloramphenicol is a broad-spectrum antimicrobic that also inhibits a range of gram-positive and gram-negative bacteria. Sheep blood is used to enrich the base and support growth of systemic fungi that may grow poorly on nonenriched medium.• Product meets ASM and CLSI reference methods and performance criteria.• BHI should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.