Learn More
S2 Media 15x100mm Bismuth Sulfite Glucose Glycine Yeast (BiGGY, Nickerson Medium) Agar, Deep Fill, 10/pack
Catalog No. 502995811
Encompass_Preferred
Bismuth Sulfite Glucose Glycine Yeast (BiGGY) Agar, also known as Nickerson Medium, is a selective and non-differential medium used for the detection, isolation, and presumptive identification of Candida species. Many strains of yeast, including Candida, reduce bismuth sulfite. During this process, significant amounts of sulfide are produced and combine with the bismuth to precipitate or impart a brown-black color to the colonies on the medium. Bismuth sulfite also serves to suppress bacterial growth.• Product pH will decrease. This is normal and does not impact performance.• Product meets CLSI reference methods and performance criteria.• SSDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5811 Supplier S2 Media Supplier No. 5068
Bismuth Sulfite Glucose Glycine Yeast (BiGGY) Agar, also known as Nickerson Medium, is a selective and non-differential medium used for the detection, isolation, and presumptive identification of Candida species. Many strains of yeast, including Candida, reduce bismuth sulfite. During this process, significant amounts of sulfide are produced and combine with the bismuth to precipitate or impart a brown-black color to the colonies on the medium. Bismuth sulfite also serves to suppress bacterial growth.• Product pH will decrease. This is normal and does not impact performance.• Product meets CLSI reference methods and performance criteria.• SSDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.