Learn More
S2 Media 15x100mm Bile Esculin Agar (BEA), 10/pack
Catalog No. 502995761
Encompass_Preferred
Bile Esculin Agar (BEA) is a selective and differential medium utilized for the isolation of enterococci and group D streptococci, including Streptococcus bovis, from non-group D streptococci. Facklam and Moody demonstrated in 1970 that using a bile esculin test was a reliable way of identifying group D streptococci. When using BEA, they determined that all group D streptococci blacken this medium. Other studies have been completed using BEA for the presumptive identification of Enterobacter spp., Klebsiella spp., and Serratia spp.• Product meets ASM, CLSI and USP-NF reference methods and performance criteria.• BEA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5761 Supplier S2 Media Supplier No. 5132
Bile Esculin Agar (BEA) is a selective and differential medium utilized for the isolation of enterococci and group D streptococci, including Streptococcus bovis, from non-group D streptococci. Facklam and Moody demonstrated in 1970 that using a bile esculin test was a reliable way of identifying group D streptococci. When using BEA, they determined that all group D streptococci blacken this medium. Other studies have been completed using BEA for the presumptive identification of Enterobacter spp., Klebsiella spp., and Serratia spp.• Product meets ASM, CLSI and USP-NF reference methods and performance criteria.• BEA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.