Learn More
S2 Media 15x100mm Bile Esculin Azide Agar with Vancomycin and Streptomycin (BEAVS), 10/pack
Catalog No. 502995904
Encompass_Preferred
Bile Esculin Azide Agar with Vancomycin and Streptomycin (BEAVS) is a selective and differential medium used for isolation and presumptive identification of group D streptococci, Enterococcus faecalis, and Enterococcus faecium. Group D streptococci and enterococci hydrolyze esculin in the presence of bile, resulting in esculetin and dextrose. Esculetin reacts with ferric ions to form brown-black colonies. Oxgall (bile salts) inhibit most strains of non-group D streptococci and enterococci. The medium is supplemented with 8µg/mL Vancomycin and 100µg/mL Streptomycin. Streptomycin inhibits gram-negative organisms present in background flora. Vancomycin aids recovery of VRE streptococci and enterococci. • Product meets CLSI reference methods and performance criteria.• BEAVS should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5904 Supplier S2 Media Supplier No. 5066
Bile Esculin Azide Agar with Vancomycin and Streptomycin (BEAVS) is a selective and differential medium used for isolation and presumptive identification of group D streptococci, Enterococcus faecalis, and Enterococcus faecium. Group D streptococci and enterococci hydrolyze esculin in the presence of bile, resulting in esculetin and dextrose. Esculetin reacts with ferric ions to form brown-black colonies. Oxgall (bile salts) inhibit most strains of non-group D streptococci and enterococci. The medium is supplemented with 8µg/mL Vancomycin and 100µg/mL Streptomycin. Streptomycin inhibits gram-negative organisms present in background flora. Vancomycin aids recovery of VRE streptococci and enterococci. • Product meets CLSI reference methods and performance criteria.• BEAVS should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.