Learn More
S2 Media 15x100mm Chameleon™ COLOREX™ Orientation and Tryptic Soy Agar with 5% Sheep Blood (Orient|BAP), Biplates, 10/pack
Catalog No. 502995785
Encompass_Preferred
Chameleon™ COLOREX™ Orientation and Tryptic Soy Agar with 5% Sheep Blood (Orient|BAP) Biplates: TSA w/ 5% Sheep Blood (Blood Agar) is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. TSA w/ 5% Sheep Blood is not the medium of choice for anaerobes. Chameleon™ COLOREX™ Orientation is a non-selective, differential, chromogenic culture medium used for the qualitative direct detection and presumptive identification of uropathogens associated with urinary tract infections (UTI). • Product complies with BAM, CLSI, and USDA reference methods and performance criteria.• Media should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5785 Supplier S2 Media Supplier No. 5453
Chameleon™ COLOREX™ Orientation and Tryptic Soy Agar with 5% Sheep Blood (Orient|BAP) Biplates: TSA w/ 5% Sheep Blood (Blood Agar) is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. TSA w/ 5% Sheep Blood is not the medium of choice for anaerobes. Chameleon™ COLOREX™ Orientation is a non-selective, differential, chromogenic culture medium used for the qualitative direct detection and presumptive identification of uropathogens associated with urinary tract infections (UTI). • Product complies with BAM, CLSI, and USDA reference methods and performance criteria.• Media should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.