Learn More
S2 Media 15x100mm Lin's Cupric Sulfate Medium Wallerstein Laboratory Differential Agar (LCSM|WLD) Biplates, 10/pack
Catalog No. 502995850
Encompass_Preferred
Lin's Cupric Sulfate Medium | Wallerstein Laboratories Differential Agar (LCSM|WLD) Biplates. LCSM is a selective and non-differential medium used for the cultivation and isolation of wild yeast species. Cupric sulfate inhibits brewer's yeast (Saccharomyces cerevisiae). WLD is a selective and differential medium used for the cultivation and isolation of bacteria. Cycloheximide inhibits fungi. Both formulations are utilized for recovery of organisms found during industrial fermentation processes. • Product meets ASBC reference methods and performance criteria.• LCSM|WLD should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.
Catalog No. 50-299-5850 Supplier S2 Media Supplier No. 5373
Lin's Cupric Sulfate Medium | Wallerstein Laboratories Differential Agar (LCSM|WLD) Biplates. LCSM is a selective and non-differential medium used for the cultivation and isolation of wild yeast species. Cupric sulfate inhibits brewer's yeast (Saccharomyces cerevisiae). WLD is a selective and differential medium used for the cultivation and isolation of bacteria. Cycloheximide inhibits fungi. Both formulations are utilized for recovery of organisms found during industrial fermentation processes. • Product meets ASBC reference methods and performance criteria.• LCSM|WLD should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.