Learn More
S2 Media 15x100mm Middlebrook 7H11 Agar | Middlebrook 7H11 Selective Agar Biplates, 10/pack
Catalog No. 502995837
Encompass_Preferred
Middlebrook 7H11 Agar | Middlebrook 7H11 Selective Agar biplates are used in qualitative procedures for the isolation and cultivation of Mycobacterium species. Middlebrook 7H11 Agar is a non-selective and non-differential culture media, which was developed in 1968 as a modification of Middlebrook 7H10 media. Middlebrook 7H11 Selective Agar is a selective and non-differential culture media, which was formulated in 1972 in order to recover mycobacteria from specimens containing mixed flora. Carbenicillin and polymyxin B inhibit most Enterobacteriaceae species, trimethoprim lactate inhibits Proteus species, and amphotericin B inhibits yeasts.• Product meets CLSI performance criteria.• Middlebrook biplates should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5837 Supplier S2 Media Supplier No. 5383
Middlebrook 7H11 Agar | Middlebrook 7H11 Selective Agar biplates are used in qualitative procedures for the isolation and cultivation of Mycobacterium species. Middlebrook 7H11 Agar is a non-selective and non-differential culture media, which was developed in 1968 as a modification of Middlebrook 7H10 media. Middlebrook 7H11 Selective Agar is a selective and non-differential culture media, which was formulated in 1972 in order to recover mycobacteria from specimens containing mixed flora. Carbenicillin and polymyxin B inhibit most Enterobacteriaceae species, trimethoprim lactate inhibits Proteus species, and amphotericin B inhibits yeasts.• Product meets CLSI performance criteria.• Middlebrook biplates should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.