Learn More
S2 Media 15x100mm Biplates: Tryptic Soy Agar with 5% Sheep Blood and MacConkey Agar (BAP|MAC), 10/pack
Catalog No. 502995748
Encompass_Preferred
Tryptic Soy Agar 5% Sheep Blood (Blood Agar) and MacConkey (MAC) Agar Biplates. Blood Agar is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. MAC is a selective and differential medium used for the detection and isolation of Gram-Negative enteric bacilli.• Product meets AOAC, ASM, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria, as applicable.• Biplates should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5748 Supplier S2 Media Supplier No. 5188
Tryptic Soy Agar 5% Sheep Blood (Blood Agar) and MacConkey (MAC) Agar Biplates. Blood Agar is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. MAC is a selective and differential medium used for the detection and isolation of Gram-Negative enteric bacilli.• Product meets AOAC, ASM, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria, as applicable.• Biplates should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.