Learn More
S2 Media 15x100mm Brain Heart Infusion (BHI) Agar, 10/pack
Catalog No. 502995794
Encompass_Preferred
Brain Heart Infusion (BHI) Agar is a non-selective and non-differential medium used for the cultivation of a wide variety of organisms. BHI is a general purpose nutritious agar base. This medium was first used for the isolation of dental pathogens. Phosphate buffer acts to neutralize the acids produced from the utilization of glucose and thus maintain viability. • Product meets AOAC, BAM, CLSI, EPA, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BHI Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5794 Supplier S2 Media Supplier No. 5032
Brain Heart Infusion (BHI) Agar is a non-selective and non-differential medium used for the cultivation of a wide variety of organisms. BHI is a general purpose nutritious agar base. This medium was first used for the isolation of dental pathogens. Phosphate buffer acts to neutralize the acids produced from the utilization of glucose and thus maintain viability. • Product meets AOAC, BAM, CLSI, EPA, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• BHI Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.