Learn More
S2 Media 15x100mm Burkholderia cepacia Selective Agar (BCSA), 10/pack
Catalog No. 502995760
Encompass_Preferred
Burkholderia cepacia Selective Agar (BCSA) is a selective medium utilized for the isolation of Burkholderia cepacia complex (Bcc). Burkholderia cepacia is a member of a the Burkholderia cepacia complex (Bcc) group, containing at least 18 closely related species. These species are known to present a significant health risk to immune compromised patients, patients on mechanical ventilation, and those suffering from underlying disease, including cystic fibrosis. Correct identification of the organism is critical to patient care. Members of the Bcc can for biofilms, which are difficult to eliminate from pharmaceutical water systems. • Product meets ASM, CLSI and USP-NF reference methods and performance criteria.• BCSA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5760 Supplier S2 Media Supplier No. 5124
Burkholderia cepacia Selective Agar (BCSA) is a selective medium utilized for the isolation of Burkholderia cepacia complex (Bcc). Burkholderia cepacia is a member of a the Burkholderia cepacia complex (Bcc) group, containing at least 18 closely related species. These species are known to present a significant health risk to immune compromised patients, patients on mechanical ventilation, and those suffering from underlying disease, including cystic fibrosis. Correct identification of the organism is critical to patient care. Members of the Bcc can for biofilms, which are difficult to eliminate from pharmaceutical water systems. • Product meets ASM, CLSI and USP-NF reference methods and performance criteria.• BCSA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.