Learn More
S2 Media 15x100mm CAMPY Blood Agar with 10% Sheep Blood, (CAMPY Blood, Blaser), 10/pack
Catalog No. 502995870
Encompass_Preferred
CAMPY Blood Agar is an enriched, selective and differential medium used for the isolation, cultivation, and presumptive identification of Campylobacter species, including Campylobacter jejuni. The base is Blaser's formulation which utilizes Brucella Agar supplemented with 10% sheep agar and five antimicrobial agents including amphotericin B, polymyxin B, cephalothin, trimethoprim, and vancomycin. The antimicrobial agents act to suppress the normal flora present in fecal specimens. (This product is equivalent to Blaser, Campylobacter Agar with 5 Antimicrobics.)• Product meets CLSI and NF reference methods and performance criteria.• CAMPY Blood Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5870 Supplier S2 Media Supplier No. 5035
CAMPY Blood Agar is an enriched, selective and differential medium used for the isolation, cultivation, and presumptive identification of Campylobacter species, including Campylobacter jejuni. The base is Blaser's formulation which utilizes Brucella Agar supplemented with 10% sheep agar and five antimicrobial agents including amphotericin B, polymyxin B, cephalothin, trimethoprim, and vancomycin. The antimicrobial agents act to suppress the normal flora present in fecal specimens. (This product is equivalent to Blaser, Campylobacter Agar with 5 Antimicrobics.)• Product meets CLSI and NF reference methods and performance criteria.• CAMPY Blood Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.