Learn More
S2 Media 15x100mm CDC Anaerobe Agar with 5% Sheep Blood (CDC Anaerobic Sheep Blood Agar), 10/pack
Catalog No. 502995830
Encompass_Preferred
CDC Anaerobe Agar with 5% Sheep Blood (CDC Anaerobic Blood Agar) is an enriched, non-selective, differential medium used for the isolation and cultivation of fastidious obligate anaerobes. The medium was formulated by Dowell et al. of the Centers for Disease Control and Prevention for the isolation and cultivation of a wide variety of obligate anaerobes found in clinical materials.• Product meets BAM and CLSI reference methods and performance criteria.• CDC Anaerobic Blood Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5830 Supplier S2 Media Supplier No. 5031
CDC Anaerobe Agar with 5% Sheep Blood (CDC Anaerobic Blood Agar) is an enriched, non-selective, differential medium used for the isolation and cultivation of fastidious obligate anaerobes. The medium was formulated by Dowell et al. of the Centers for Disease Control and Prevention for the isolation and cultivation of a wide variety of obligate anaerobes found in clinical materials.• Product meets BAM and CLSI reference methods and performance criteria.• CDC Anaerobic Blood Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.