Learn More
S2 Media 15x100mm Chocolate Agar, 10/pack
Catalog No. 502995900
Encompass_Preferred
Chocolate Agar is a non-selective, differential medium utilized for the isolation and cultivation of fastidious microorganisms, such as Neisseria and Haemophilus species. (This product demonstrates equivalent performance to Chocolate II formulations in both growth recovery and rate (time).) • Product meets CLSI performance criteria.• Chocolate Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5900 Supplier S2 Media Supplier No. 5012
Chocolate Agar is a non-selective, differential medium utilized for the isolation and cultivation of fastidious microorganisms, such as Neisseria and Haemophilus species. (This product demonstrates equivalent performance to Chocolate II formulations in both growth recovery and rate (time).) • Product meets CLSI performance criteria.• Chocolate Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.