Learn More
S2 Media 15x100mm Corynebacterium kroppenstedtii Selective Media (CKSM) Agar, 10/pack
Catalog No. 502995847
Encompass_Preferred
Corynebacterium kroppenstedtii Selective Media (CKSM) Agar is a selective and differential agar used for the cultivation and isolation of C. kroppenstedtii from clinical specimens. • Product meets CLSI reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• CKSM should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5847 Supplier S2 Media Supplier No. 5367
Corynebacterium kroppenstedtii Selective Media (CKSM) Agar is a selective and differential agar used for the cultivation and isolation of C. kroppenstedtii from clinical specimens. • Product meets CLSI reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• CKSM should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.